siRNA Details
| Gene Symbol: ZNF845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321522.1 | NP_001308451.1 | 5 | 3746 | ||
| NM_001321523.1 | NP_001308452.1 | 5 | 3685 | |||
| NM_001321524.1 | NP_001308453.1 | 5 | 3693 | |||
| NM_138374.2 | NP_612383.1 | 4 | 3591 | |||
| GenBank mRNA | AK295079.1 | - | 6 | 3413 | ||
| BC007307.1 | - | 1 | 2447 | |||

ZNF845
Genomic Map