siRNA Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127181.2 | NP_001120653.1 | 7 | 1731 | ||
| NM_001171182.1 | NP_001164653.1 | 5 | 1204 | |||
| NM_033319.3 | NP_201576.1 | 6 | 1593 | |||
| GenBank mRNA | BC007071.1 | - | 6 | 1128 | 3682908 | |
| BC019022.2 | - | 5 | 1205 | |||
| BC033154.1 | - | 7 | 1414 | 4139038 | ||
| BC066658.1 | - | 6 | 1275 | 6172418 | ||

CENPL
Genomic Map