siRNA Details
| Gene Symbol: ZNF804A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194250.1 | NP_919226.1 | 4 | 1965 | ||
| GenBank mRNA | AK127762.1 | - | 3 | 1139 | ||
| AK131402.1 | - | 4 | 1805 | |||
| AK307596.1 | - | 4 | 1911 | |||
| AY327407.1 | - | 4 | 1965 | |||
| BC136602.1 | - | 4 | 1620 | 9020592 | ||
| BC142612.1 | - | 4 | 1765 | |||
| BC150198.1 | - | 4 | 1822 | 40148778 | ||

ZNF804A
Genomic Map