siRNA Details
| Gene Symbol: MEX3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093725.1 | NP_001087194.1 | 2 | 879 | ||
| GenBank mRNA | AK125482.1 | - | 1 | 381 | ||
| AY950677.1 | - | 2 | 879 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEX3A| Gene Symbol: MEX3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093725.1 | NP_001087194.1 | 2 | 879 | ||
| GenBank mRNA | AK125482.1 | - | 1 | 381 | ||
| AY950677.1 | - | 2 | 879 | |||