siRNA Details
| Gene Symbol: EGFEM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021485.2 | 13 | 2052 | |||
| GenBank mRNA | AF086185.1 | - | 1 | 168 | ||
| BC011266.2 | - | 3 | 348 | |||
| BC103698.1 | - | 1 | 168 | |||
| BC104748.1 | - | 1 | 168 | |||
| BC105690.1 | - | 1 | 168 | |||

EGFEM1P
Genomic Map