siRNA Details
Gene Symbol: LOC93622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026375.1 | - | 1 | 860 | ||
BC021542.1 | - | 1 | 832 | |||
BC066911.1 | - | 1 | 940 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC93622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026375.1 | - | 1 | 860 | ||
BC021542.1 | - | 1 | 832 | |||
BC066911.1 | - | 1 | 940 |