siRNA Details
| Gene Symbol: ST7-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002331.3 | 1 | 80 | |||
| NR_109981.1 | 1 | 80 | ||||
| GenBank mRNA | AF400040.1 | - | LIT1999 ST7OT2 |
1 | 46 | |
| AF400043.1 | - | LIT1938 ST7OT2 |
2 | 444 | ||
| AF400045.1 | - | LIT1899 ST7OT2 |
1 | 71 | ||
| DB054726.1 | - | 1 | 80 | |||

ST7-AS2
Genomic Map