siRNA Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080457.1 | NP_001073926.1 | 3 | 1885 | ||
| XM_005259429.4 | XP_005259486.1 | 3 | 2064 | |||
| XM_006723506.3 | XP_006723569.1 | 4 | 2166 | |||
| XM_011527520.2 | XP_011525822.1 | 3 | 3312 | |||
| GenBank mRNA | AL137451.1 | - | 1 | 1105 | ||
| BC019687.3 | - | 1 | 1129 | |||
| BC032460.1 | - | 2 | 1760 | |||
| BC056207.1 | - | 1 | 92 | |||

LRRC4B
Genomic Map