siRNA Details
| Gene Symbol: LENG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301782.1 | NP_001288711.1 | 1 | 1557 | ||
| NM_198988.1 | NP_945339.2 | 1 | 1560 | |||
| GenBank mRNA | BC015921.1 | - | 1 | 1560 | 4551151 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LENG9| Gene Symbol: LENG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301782.1 | NP_001288711.1 | 1 | 1557 | ||
| NM_198988.1 | NP_945339.2 | 1 | 1560 | |||
| GenBank mRNA | BC015921.1 | - | 1 | 1560 | 4551151 | |