siRNA Details
| Gene Symbol: HSPB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033194.2 | NP_149971.1 | 1 | 344 | ||
| GenBank mRNA | AK093488.1 | - | 1 | 1652 | ||
| AK130947.1 | - | 1 | 252 | |||
| BC093991.1 | - | 1 | 265 | 7939836 | ||
| BC093993.1 | - | 1 | 265 | 7939838 | ||

HSPB9
Genomic Map