siRNA Details
| Gene Symbol: PHF21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135862.2 | NP_001129334.1 | 13 | 1291 | ||
| NM_001242450.1 | NP_001229379.1 | 12 | 1331 | |||
| NM_001284296.1 | NP_001271225.1 | 13 | 1377 | |||
| NM_138415.4 | NP_612424.1 | 12 | 1462 | |||
| NR_134535.1 | 11 | 1223 | ||||
| XM_006724122.1 | XP_006724185.1 | 12 | 1471 | |||
| GenBank mRNA | AK091480.1 | - | 13 | 1377 | ||
| AK092243.1 | - | 13 | 1291 | |||
| AK095777.1 | - | 6 | 443 | |||
| AK297317.1 | - | 12 | 1324 | |||
| BC012187.1 | - | 12 | 1462 | 4634631 | ||
| CR456448.1 | - | 12 | 1218 | |||
| KU178842.1 | - | 12 | 1311 | |||
| KU178843.1 | - | 12 | 1185 | |||
| KU178844.1 | - | 11 | 1162 | |||

PHF21B
Genomic Map