siRNA Details
| Gene Symbol: C12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301834.1 | NP_001288763.1 | 3 | 359 | ||
| NM_001301836.1 | NP_001288765.1 | 2 | 356 | |||
| NM_001301838.1 | NP_001288767.1 | 2 | 584 | |||
| NM_138425.3 | NP_612434.1 | 2 | 488 | |||
| GenBank mRNA | AK310266.1 | - | 3 | 519 | ||
| AK311912.1 | - | 2 | 286 | |||
| BC009925.2 | - | 2 | 265 | 3537206 | ||
| BE888740.1 | - | 2 | 275 | |||
| BM781587.1 | - | 2 | 488 | |||
| BP318750.2 | - | 92 | ||||
| CB161655.1 | - | 3 | 359 | |||
| DB494917.1 | - | 2 | 356 | |||

C12orf57
Genomic Map