siRNA Details
| Gene Symbol: C12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301834.1 | NP_001288763.1 | 4 | 491 | ||
| NM_001301836.1 | NP_001288765.1 | 3 | 488 | |||
| NM_001301837.1 | NP_001288766.1 | 3 | 533 | |||
| NM_001301838.1 | NP_001288767.1 | 3 | 716 | |||
| NM_138425.3 | NP_612434.1 | 3 | 620 | |||
| NR_126035.1 | 3 | 629 | ||||
| GenBank mRNA | AK311912.1 | - | 3 | 418 | ||
| AV751928.1 | - | 371 | ||||
| BC009925.2 | - | 3 | 397 | 3537206 | ||
| BE888740.1 | - | 3 | 407 | |||
| BM687523.1 | - | 3 | 520 | |||
| BP318750.2 | - | 224 | ||||
| CB161655.1 | - | 4 | 491 | |||

C12orf57
Genomic Map