siRNA Details
| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 2 | 1626 | |||
| GenBank mRNA | AK096901.1 | - | 2 | 1331 | ||
| AK097739.1 | - | 2 | 894 | |||
| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003608.1 | 3 | 779 | |||
| GenBank mRNA | AK057257.1 | - | 3 | 731 | ||
| AK315686.1 | - | 3 | 727 | |||
| BC009991.2 | - | 3 | 633 | |||

THAP7-AS1
Genomic Map