siRNA Details
| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003608.1 | 3 | 1024 | |||
| GenBank mRNA | AK057257.1 | - | 3 | 976 | ||
| AK315686.1 | - | 3 | 972 | |||
| BC009991.2 | - | 3 | 878 | |||
| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 2 | 1381 | |||
| GenBank mRNA | AK096901.1 | - | 2 | 1086 | ||
| AK097739.1 | - | 2 | 649 | |||

TUBA3FP
Genomic Map