siRNA Details
| Gene Symbol: TTTY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001524.1 | 3 | 295 | |||
| Gene Symbol: TTTY3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002176.1 | 3 | 295 | |||

TTTY3
Genomic Map