siRNA Details
| Gene Symbol: PSMG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021487.2 | 5 | 1955 | |||
| NR_027328.1 | 4 | 1593 | ||||
| NR_027329.1 | 5 | 1701 | ||||
| GenBank mRNA | AB067495.1 | - | 4 | 1571 | ||
| AK054665.1 | - | 4 | 1593 | |||
| BX538028.1 | - | 5 | 1639 | |||

PSMG3-AS1
Genomic Map