siRNA Details
| Gene Symbol: CCDC85A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080433.1 | NP_001073902.1 | 2 | 819 | ||
| XM_005264125.1 | XP_005264182.1 | 2 | 344 | |||
| GenBank mRNA | AB067499.2 | - | 2 | 903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC85A| Gene Symbol: CCDC85A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080433.1 | NP_001073902.1 | 2 | 819 | ||
| XM_005264125.1 | XP_005264182.1 | 2 | 344 | |||
| GenBank mRNA | AB067499.2 | - | 2 | 903 | ||