siRNA Details
| Gene Symbol: MYSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085487.2 | NP_001078956.1 | 8 | 539 | ||
| XM_006710314.3 | XP_006710377.1 | 8 | 555 | |||
| XM_011540573.2 | XP_011538875.1 | 9 | 519 | |||
| XM_011540574.2 | XP_011538876.1 | 8 | 787 | |||
| XM_017000195.1 | XP_016855684.1 | 9 | 591 | |||
| XM_017000196.1 | XP_016855685.1 | 6 | 441 | |||
| XM_017000197.1 | XP_016855686.1 | 4 | 219 | |||
| GenBank mRNA | AB067502.1 | - | 4 | 565 | ||
| AK126835.1 | - | 2 | 88 | |||
| BX648168.1 | - | 5 | 275 | |||

MYSM1
Genomic Map