siRNA Details
| Gene Symbol: RNF157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052916.2 | NP_443148.1 | 2 | 243 | ||
| XM_005257007.4 | XP_005257064.1 | 2 | 302 | |||
| XM_011524273.2 | XP_011522575.1 | 2 | 302 | |||
| XM_017024117.1 | XP_016879606.1 | 2 | 302 | |||
| XM_017024118.1 | XP_016879607.1 | 2 | 302 | |||
| XM_017024119.1 | XP_016879608.1 | 2 | 302 | |||
| XM_017024120.1 | XP_016879609.1 | 1 | 33 | |||
| GenBank mRNA | AB067504.2 | - | 2 | 243 | ||
| AK091467.1 | - | 2 | 126 | |||

RNF157
Genomic Map