siRNA Details
| Gene Symbol: RBM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005105.4 | NP_005096.1 | 6 | 3844 | ||
| GenBank mRNA | AL049219.1 | - | 1 | 1233 | ||
| BC071577.1 | - | 6 | 3792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBM8A| Gene Symbol: RBM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005105.4 | NP_005096.1 | 6 | 3844 | ||
| GenBank mRNA | AL049219.1 | - | 1 | 1233 | ||
| BC071577.1 | - | 6 | 3792 | |||