siRNA Details
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037844.2 | 7 | 4569 | |||
GenBank mRNA | AI469921.1 | - | 1 | 54 | ||
AK021767.1 | - | 3 | 1579 | |||
AK128532.1 | - | 6 | 3472 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037844.2 | 7 | 4569 | |||
GenBank mRNA | AI469921.1 | - | 1 | 54 | ||
AK021767.1 | - | 3 | 1579 | |||
AK128532.1 | - | 6 | 3472 |