siRNA Details
| Gene Symbol: KCTD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138444.3 | NP_612453.1 | 1 | 967 | ||
| GenBank mRNA | AF359381.1 | - | 1 | 952 | ||
| AK128363.1 | - | 1 | 970 | |||
| BC013764.1 | - | 1 | 797 | 4521096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD12| Gene Symbol: KCTD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138444.3 | NP_612453.1 | 1 | 967 | ||
| GenBank mRNA | AF359381.1 | - | 1 | 952 | ||
| AK128363.1 | - | 1 | 970 | |||
| BC013764.1 | - | 1 | 797 | 4521096 | ||