siRNA Details
| Gene Symbol: RAB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193532.1 | NP_001180461.1 | 2 | 486 | ||
| NM_152304.1 | NP_689517.1 | 2 | 436 | |||
| XM_017000226.1 | XP_016855715.1 | 2 | 507 | |||
| GenBank mRNA | AK312176.1 | - | 2 | 373 | ||
| BC033175.1 | - | 2 | 436 | 4561064 | ||
| CX788289.1 | - | 2 | 493 | |||

RAB42
Genomic Map