siRNA Details
| Gene Symbol: ZNF501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 1186 | ||
| NM_145044.3 | NP_659481.2 | 3 | 893 | |||
| GenBank mRNA | AK297216.1 | - | 3 | 875 | ||
| AK316574.1 | - | 3 | 893 | |||
| BC013762.1 | - | 3 | 868 | 4521303 | ||

ZNF501
Genomic Map