siRNA Details
| Gene Symbol: ZNF493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076678.2 | NP_001070146.1 | 3 | 350 | ||
| NM_145326.2 | NP_663299.2 | 3 | 353 | |||
| GenBank mRNA | AK092178.1 | - | 3 | 336 | ||
| BC006408.1 | - | 3 | 316 | |||
| BX648842.1 | - | 3 | 340 | |||
| DA804007.1 | - | 3 | 353 | |||

ZNF493
Genomic Map