siRNA Details
| Gene Symbol: NXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138454.1 | NP_612463.1 | 1 | 319 | ||
| GenBank mRNA | BC014127.1 | - | 1 | 319 | 4579939 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NXNL1| Gene Symbol: NXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138454.1 | NP_612463.1 | 1 | 319 | ||
| GenBank mRNA | BC014127.1 | - | 1 | 319 | 4579939 | |