siRNA Details
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 7, 8 | 897 | ||
GenBank mRNA | AK289402.1 | - | 7, 8 | 884 | ||
AK315729.1 | - | 7, 8 | 894 | |||
AY358952.1 | - | 7, 8 | 1015 | |||
BC014320.1 | - | 7, 8 | 901 | 3686673 |
