siRNA Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053050.4 | NP_444278.1 | 2, 3 | 368 | ||
| GenBank mRNA | BC012163.1 | - | 2, 3 | 219 | 4622995 | |
| BG196300.1 | - | 2, 3 | 353 | |||
| BQ014848.1 | - | 1, 2 | 274 | |||
| BQ688110.1 | - | 2, 3 | 225 | |||

MRPL53
Genomic Map