siRNA Details
| Gene Symbol: CATSPER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282309.1 | NP_001269238.1 | 4 | 300 | ||
| NM_001282310.1 | NP_001269239.1 | 3 | 215 | |||
| NM_054020.3 | NP_473361.1 | 3 | 401 | |||
| NM_172095.2 | NP_742093.1 | 3 | 401 | |||
| NR_110319.1 | 2 | 188 | ||||
| XM_017021904.1 | XP_016877393.1 | 3 | 407 | |||
| GenBank mRNA | AF411817.1 | - | 2 | 186 | ||
| AF411818.1 | - | 2 | 186 | |||
| AF411819.1 | - | 2 | 186 | |||
| AK122607.1 | - | 4 | 299 | |||
| BC028728.1 | - | 3 | 406 | |||
| BC064387.1 | - | 3 | 215 | 5744415 | ||
| BX648099.1 | - | 3 | 437 | |||
| DQ655952.2 | - | 3 | 213 | |||

CATSPER2
Genomic Map