siRNA Details
| Gene Symbol: RLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001311197.1 | NP_001298126.1 | 1 | 72 | ||
| NM_080864.3 | NP_543140.1 | 1 | 72 | |||
| GenBank mRNA | AB076563.1 | - | 1 | 15 | ||
| AF447451.1 | - | 1 | 15 | |||
| AY358181.1 | - | 2 | 855 | |||
| BC140935.1 | - | 1 | 72 | 9056747 | ||

RLN3
Genomic Map