siRNA Details
| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 2 | 677 | |||
| NR_026553.1 | 2 | 321 | ||||
| GenBank mRNA | AW451670.1 | - | 2 | 512 | ||
| AY063458.1 | - | 2 | 677 | |||
| AY063459.1 | - | 2 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00161| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 2 | 677 | |||
| NR_026553.1 | 2 | 321 | ||||
| GenBank mRNA | AW451670.1 | - | 2 | 512 | ||
| AY063458.1 | - | 2 | 677 | |||
| AY063459.1 | - | 2 | 321 | |||