siRNA Details
| Gene Symbol: FAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008781.2 | NP_001008781.2 | 1 | 2329 | ||
| XM_017017178.1 | XP_016872667.1 | 2 | 2445 | |||
| XM_017017179.1 | XP_016872668.1 | 3 | 2667 | |||
| XM_017017180.1 | XP_016872669.1 | 2 | 2444 | |||
| XM_017017181.1 | XP_016872670.1 | 2 | 2444 | |||
| XM_017017182.1 | XP_016872671.1 | 2 | 2444 | |||
| XM_017017183.1 | XP_016872672.1 | 2 | 2444 | |||
| XM_017017184.1 | XP_016872673.1 | 2 | 2443 | |||
| GenBank mRNA | AK054903.1 | - | 1 | 1061 | ||

FAT3
Genomic Map