siRNA Details
| Gene Symbol: C14orf79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174891.3 | NP_777551.2 | 4 | 911 | ||
| XM_005267317.3 | XP_005267374.1 | 4 | 1389 | |||
| XM_005267318.3 | XP_005267375.1 | 4 | 1389 | |||
| GenBank mRNA | AF038188.1 | - | 4 | 475 | ||
| AK057249.1 | - | 5 | 987 | |||
| BC003539.1 | - | 3 | 223 | |||
| BC011537.1 | - | 3 | 741 | |||
| BC025175.1 | - | 4 | 422 | |||
| BC137495.1 | - | 4 | 854 | 9021495 | ||
| BC144671.1 | - | 4 | 840 | |||

C14orf79
Genomic Map