siRNA Details
| Gene Symbol: SLC51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178859.3 | NP_849190.2 | 2, 3 | 418 | ||
| XM_005254159.4 | XP_005254216.1 | 2, 3 | 393 | |||
| GenBank mRNA | AI793107.1 | - | 2, 3 | 418 | ||
| AK129601.1 | - | 2, 3 | 321 | |||
| AY194242.1 | - | 1, 2 | 127 | |||
| BC103842.1 | - | 1, 2 | 124 | 40002450 | ||
| BC103843.1 | - | 1, 2 | 124 | 40002452 | ||
| BC103844.1 | - | 1, 2 | 124 | 40002453 | ||

SLC51B
Genomic Map