siRNA Details
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255859.4 | XP_005255916.3 | 1 | 1455 | ||
XM_005255860.3 | XP_005255917.3 | 1 | 1462 | |||
XM_005255861.4 | XP_005255918.3 | 1 | 1455 | |||
XM_017023084.1 | XP_016878573.1 | 1 | 1627 |
