siRNA Details
| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 1 | 280 | |||
| GenBank mRNA | AK096394.1 | - | 1 | 314 | ||
| AK291606.1 | - | 1 | 264 | |||
| BC062576.1 | - | 1 | 246 | |||
| BC080650.1 | - | 1 | 210 | |||
| BC093998.1 | - | 1 | 280 | |||
| BC105289.1 | - | 1 | 221 | |||
| BC111952.1 | - | 1 | 280 | |||

LOC652276
Genomic Map