siRNA Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144972.1 | NP_001138444.1 | 3 | 408 | ||
| NM_153337.2 | NP_699168.1 | 3 | 408 | |||
| NM_182854.3 | NP_878274.1 | 3 | 408 | |||
| GenBank mRNA | AK055837.1 | - | 3 | 373 | ||
| AK292650.1 | - | 3 | 367 | |||
| AK292705.1 | - | 3 | 373 | |||
| AY302441.1 | - | 3 | 310 | |||
| BC027944.1 | - | 3 | 357 | |||
| BC063423.1 | - | 3 | 349 | |||
| BC119626.1 | - | 3 | 261 | |||

SNX20
Genomic Map