siRNA Details
Gene Symbol: FLJ40194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034161.1 | 2 | 426 | |||
GenBank mRNA | AK097513.1 | - | 2 | 430 | ||
BI463223.1 | - | 2 | 431 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ40194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034161.1 | 2 | 426 | |||
GenBank mRNA | AK097513.1 | - | 2 | 430 | ||
BI463223.1 | - | 2 | 431 |