siRNA Details
| Gene Symbol: MIEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144900.1 | NP_001138372.1 | 2, 3 | 352 | ||
| NM_139162.3 | NP_631901.2 | 2, 3 | 352 | |||
| NM_148886.1 | NP_683684.2 | 2, 3 | 200 | |||
| XM_017024190.1 | XP_016879679.1 | 2, 3 | 697 | |||
| GenBank mRNA | AF467443.1 | - | 2, 3 | 200 | ||
| AK056165.1 | - | 2, 3 | 331 | |||
| AK128310.1 | - | 2, 3 | 255 | |||
| BC014973.1 | - | 2, 3 | 200 | 4859126 | ||
| DB183618.1 | - | 2, 3 | 358 | |||

MIEF2
Genomic Map