siRNA Details
| Gene Symbol: C19orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308240.1 | NP_001295169.1 | 5 | 945 | ||
| NM_205767.2 | NP_991330.1 | 4 | 749 | |||
| XM_017026248.1 | XP_016881737.1 | 4 | 690 | |||
| GenBank mRNA | AI300708.1 | - | 1 | 154 | ||
| AJ707830.1 | - | 4 | 495 | |||
| BC009557.1 | - | 4 | 383 | 3899541 | ||
| BC042386.1 | - | 4 | 358 | 5288954 | ||

C19orf70
Genomic Map