siRNA Details
| Gene Symbol: ZNF813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 1565 | ||
| GenBank mRNA | AK091460.1 | - | 1 | 713 | ||
| AK131422.1 | - | 1 | 948 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF813| Gene Symbol: ZNF813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 1565 | ||
| GenBank mRNA | AK091460.1 | - | 1 | 713 | ||
| AK131422.1 | - | 1 | 948 | |||