siRNA Details
| Gene Symbol: ZNF787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002836.3 | NP_001002836.2 | 2 | 256 | ||
| XM_011526445.2 | XP_011524747.1 | 2 | 1940 | |||
| GenBank mRNA | AF000560.1 | - | 1 | 53 | ||
| BC012495.1 | - | 2 | 160 | |||
| BC077728.1 | - | 2 | 163 | 6210543 | ||

ZNF787
Genomic Map