siRNA Details
| Gene Symbol: PLK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243079.1 | NP_001230008.1 | 7, 8 | 908 | ||
| GenBank mRNA | AK054808.1 | - | 7, 8 | 908 | ||
| AK122648.1 | - | 6, 7 | 1080 | |||
| BC100791.2 | - | 4, 5 | 472 | |||
| BC100792.1 | - | 4, 5 | 472 | |||
| DQ424898.1 | - | 1, 2 | 192 | |||

PLK5
Genomic Map