siRNA Details
| Gene Symbol: RPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122965.1 | NP_001116437.1 | 3 | 2255 | ||
| GenBank mRNA | AK096436.1 | - | 1 | 1165 | ||
| AY396742.1 | - | 3 | 2255 | |||
| BC112216.1 | - | 1 | 123 | |||
| BC112218.1 | - | 1 | 122 | |||

RPTN
Genomic Map