siRNA Details
| Gene Symbol: CCDC163P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033296.1 | 3 | 689 | |||
| NR_033298.1 | 3 | 689 | ||||
| GenBank mRNA | AK300867.1 | - | 3 | 579 | ||
| BC036618.2 | - | 2 | 222 | |||
| BC047421.1 | - | 3 | 658 | |||
| BC101255.2 | - | 3 | 584 | |||
| BC101256.1 | - | 3 | 584 | |||
| BC101257.1 | - | 3 | 584 | |||
| BC113060.1 | - | 3 | 584 | |||
| BC113061.1 | - | 3 | 584 | |||

CCDC163P
Genomic Map