siRNA Details
| Gene Symbol: C1orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122961.1 | NP_001116433.1 | 2 | 278 | ||
| XM_006710342.3 | XP_006710405.1 | 2 | 624 | |||
| XM_011540647.2 | XP_011538949.1 | 2 | 655 | |||
| XM_011540648.1 | XP_011538950.1 | 2 | 660 | |||
| GenBank mRNA | BG772797.1 | - | 2 | 278 | ||

C1orf194
Genomic Map