siRNA Details
| Gene Symbol: C1orf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142726.1 | NP_001136198.1 | 2, 3 | 709 | ||
| NM_198446.2 | NP_940848.2 | 2, 3 | 842 | |||
| GenBank mRNA | AK127381.1 | - | 2, 3 | 346 | ||
| AY517499.1 | - | 2, 3 | 497 | |||
| BC016402.1 | - | 1, 2 | 1755 | |||
| BC057819.1 | - | 2, 3 | 347 | 4792982 | ||
| BC107856.1 | - | 1, 2 | 457 | |||
| CV575592.1 | - | 2, 3 | 415 | |||

C1orf122
Genomic Map