siRNA Details
| Gene Symbol: C1orf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142726.1 | NP_001136198.1 | 3 | 763 | ||
| NM_198446.2 | NP_940848.2 | 3 | 896 | |||
| GenBank mRNA | AK127381.1 | - | 3 | 400 | ||
| AY517499.1 | - | 3 | 551 | |||
| BC016402.1 | - | 2 | 1809 | |||
| BC107856.1 | - | 2 | 511 | |||
| CV575592.1 | - | 3 | 469 | |||

C1orf122
Genomic Map