siRNA Details
| Gene Symbol: CST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008693.2 | NP_001008693.2 | 1, 2 | 361 | ||
| GenBank mRNA | AF494536.1 | - | 1, 2 | 264 | ||
| BC137302.1 | - | 1, 2 | 361 | 9021299 | ||
| BC137303.1 | - | 1, 2 | 361 | 9021300 | ||

CST9
Genomic Map